Principaux documents
SLW1060/21D
Pyrex® Watch glasses
nominal diameter 200 mm
Synonyme(s) :
Pyrex, SciLabware 1060/21D
10 EA
485.00 CHF
485.00 CHF
Date d'expédition estimée le10 avril 2025
Changer de vue
10 EA
485.00 CHF
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
485.00 CHF
Date d'expédition estimée le10 avril 2025
Conditionnement
pkg of 10
Fabricant/nom de marque
1060/21D
SciLabware
Diamètre nominal
200 mm
Description générale
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Caractéristiques et avantages
• Complies with DIN 12341• Manufactured from borosilicate glass 3.3• Heat resistant• Fire-polished edges to reduce chipping and breakage
Informations légales
PYREX is a registered trademark of Corning, Inc.