Principaux documents
PL800GH-8-6
Plas-Labs Hypalon glove box gloves
for 1 piece glove set, size 8 (for 6 in. ports)
1 PAIR
535.00 CHF
535.00 CHF
Check Cart for Availability
Changer de vue
1 PAIR
535.00 CHF
About This Item
Code UNSPSC :
46181504
Nomenclature NACRES :
NB.35
535.00 CHF
Check Cart for Availability
Matériaux
Hypalon®
Taille
8 (for 6 in. ports)
Caractéristiques
reusable
Conditionnement
pack of 1 pair
Fabricant/nom de marque
Plas-Labs 800GH-8-6
Description générale
Plas-Labs white Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Informations légales
HaackerCalc is a trademark of ChemSW, Inc.
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company