Principaux documents
NOR5X3INSV
Optimizer insert for spinner turbines
NMR tube size 3 mm , into 5 mm Varian
1 EA
154.00 CHF
154.00 CHF
Check Cart for Availability
Changer de vue
1 EA
154.00 CHF
About This Item
Code UNSPSC :
41121705
Nomenclature NACRES :
SB.32
154.00 CHF
Check Cart for Availability
Matériaux
(acetal homopolymer resin)
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Norell 5X3INS-V
Taille du tube de RMN
3 mm , into 5 mm Varian
Description générale
Precision adapter holds 3 mm, 4 mm, and 4.25 mm NMR tube in a 5 mm spinner turbine. Does not include spinner turbine.