Principaux documents
MT30034472
Mettler Toledo SevenExcellence pH/mV expansion unit
1 EA
1 250.00 CHF
1 250.00 CHF
Date d'expédition estimée le14 avril 2025
Changer de vue
1 EA
1 250.00 CHF
About This Item
Code UNSPSC :
41115603
Nomenclature NACRES :
NB.61
1 250.00 CHF
Date d'expédition estimée le14 avril 2025
Fabricant/nom de marque
Mettler-Toledo 30034472
Description générale
Can be used to expand any SevenExcellence meter to measure pH/mV.
SevenExcellence fits three modules in any composition and order.
It offers the highest possible
flexibility as you can expand the instrument at any time, adding new measurement parameters.
Informations légales
SevenExcellence is a trademark of Mettler-Toledo, Inc.