This cap is suitable for use up to 135°C and may be repeatedly autoclaved at 121°C at 15 psig for 20 minutes. To prevent damage to containers, caps should be loose during autoclaving to allow for venting.
DWK73808-33430
Kimble® Closed Top Polypropylene Screw Thread Cap With Welded Ptfe-Faced Silicone Liner
33-430
128.00 CHF
About This Item
128.00 CHF
Produits recommandés
Matériaux
PTFE/silicone liner
black polypropylene screw cap (33-430 thread)
Conditionnement
pkg of 1 packs (48 caps)
Fabricant/nom de marque
Kimble® 73808-33430
Raccords
thread
Application
Caractéristiques et avantages
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Closures are ideal for repeated autoclaving
- Welded liner technology eliminates the possibility of glue contamination
-
What is the heat capacity of these lids? Can they withstand autoclave temperatures/pressure?