Principaux documents
DWK217314401
DURAN® wide mouth reaction flask with beaded rim
round bottom glass flask, flask capacity (500 mL), single-neck
Synonyme(s) :
217314401
10 EACH
148.00 CHF
148.00 CHF
Check Cart for Availability
Changer de vue
10 EACH
148.00 CHF
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.17
148.00 CHF
Check Cart for Availability
Matériaux
round bottom glass flask
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 217314401
Contenance du flacon/de la fiole
(500 mL)
Description générale
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
Informations légales
DURAN is a registered trademark of DWK Life Sciences