Principaux documents
CLS4450500
Pyrex® baffled Erlenmeyer flask
capacity 500 mL
Synonyme(s) :
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, laboratory flasks, Corning® 4450
About This Item
Code UNSPSC :
41122104
eCl@ss :
32040501
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
flat bottom flask
narrow-mouth
narrow-mouth neck
Stérilité
non-sterile
Caractéristiques
cap: no
closure type Rubber stopper
Conditionnement
case of 6
Fabricant/nom de marque
Corning 4450-500
Capacité
500 mL
Description générale
  • Pyrex® baffled Erlenmeyer flasks feature a heavy tooled rim for maximum durability.
  • Baffle-designed Delong flasks promote optimal and consistent solution agitation when used with flask shakers.
  • Uniform wall thickness ensures an optimal balance between mechanical strength and thermal shock resistance.
  • Includes a marking spot for convenience.
Autres remarques
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.