Principaux documents
CAD7921
Animal feeding needle, non-sterile, reusable, type 304 SS
straight, size 20G, L × diam. 1 in. × 2.25 mm , ball
12 EA
1 030.00 CHF
1 030.00 CHF
Check Cart for Availability
Changer de vue
12 EA
1 030.00 CHF
About This Item
Code UNSPSC :
42142609
Nomenclature NACRES :
NB.03
1 030.00 CHF
Check Cart for Availability
Taille
20G
Conditionnement
pkg of 12 ea
L × diam.
1 in. × 2.25 mm , ball
Catégories apparentées
Caractéristiques et avantages
  • Fixed shape from stainless steel tubing
  • Stainless steel balls
  • Can be cleaned and reused as needed
  • Non-sterile