Principaux documents
CAD4190
Stainless steel 304 syringe needle
gauge 19, L 4 in.
12 EA
463.00 CHF
463.00 CHF
Check Cart for Availability
Changer de vue
12 EA
463.00 CHF
About This Item
Code UNSPSC :
42142600
Nomenclature NACRES :
NB.03
463.00 CHF
Check Cart for Availability
Description
Standard Hub with 304 Stainless Steel Tubing
Conditionnement
pack of 12 ea
Fabricant/nom de marque
Cadence Science 4190
Longueur
4 in.
Calibre
19
Catégories apparentées
Description générale
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing