Principaux documents
BR90663
BRAND® glass beaker with spout, tall form
volume 2000 mL, with graduation
Synonyme(s) :
Beakers, Glass
10 EA
406.00 CHF
406.00 CHF
Check Cart for Availability
Changer de vue
10 EA
406.00 CHF
About This Item
Code UNSPSC :
41121803
Nomenclature NACRES :
NB.43
406.00 CHF
Check Cart for Availability
Matériaux
borosilicate glass 3.3
Caractéristiques
with graduation
Fabricant/nom de marque
BRAND 90663
Volume
2000 mL
Description générale
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Autres remarques
Browse all glass beakers
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG