Principaux documents
BR61605
BRAND® silicone paste UNISILKON TK M 1011 tube of 50 g
1 EA
36.30 CHF
36.30 CHF
Check Cart for Availability
Changer de vue
1 EA
36.30 CHF
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
36.30 CHF
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 61605
Catégories apparentées
Description générale
The BRAND® silicone paste UNISILKON TK M 1011 comes in a convenient 50 g tube and features a low viscosity formulation, making it ideal for various applications. With an operating temperature range of -40 to 160 °C, this silicone paste is highly versatile and suitable for use in extreme conditions. It is specifically designed for greasing ground joints, ensuring a smooth and effective seal.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG