BR4630151
BRAND® Dispensette® S Organic, Analog-adjustable bottle-top dispenser
volume range 2.5-25 mL, with recirculation valve
Synonyme(s) :
bottle top dispensers
527.00 CHF
About This Item
527.00 CHF
Produits recommandés
Caractéristiques
incremental 0.5 mL
DE-M marking
with recirculation valve
Fabricant/nom de marque
BRAND 4630151
Gamme de volumes
2.5-25 mL
Raccords
(GL 32-33)
GL 38
S 40
Catégories apparentées
Description générale
Caractéristiques et avantages
- Extremely easy filling and dispensing
- Simple priming by optimized flow channels
- Rapid and safe volume adjustment with internal scalloped track
Les clients ont également consulté
Protocoles
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenu apparenté
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.