Principaux documents
BR150904
BRAND® watch glass
PTFE, diam. 40 mm
Synonyme(s) :
Dishes, PTFE, Watch glasses
1 EA
9.90 CHF
9.90 CHF
Check Cart for Availability
Changer de vue
1 EA
9.90 CHF
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
9.90 CHF
Check Cart for Availability
Matériaux
PTFE
Fabricant/nom de marque
BRAND 150904
Diamètre
40 mm
Catégories apparentées
Description générale
The BRAND® watch glass is made from high-quality PTFE, making it ideal for covering PTFE beakers. Its excellent chemical resistance ensures durability and reliability when handling a wide range of substances. This watch glass provides a secure cover to prevent contamination and evaporation during experiments. Its lightweight and non-reactive properties make it a valuable accessory in laboratory settings, enhancing the efficiency of various applications. This versatile tool is essential for maintaining the integrity of samples and solutions in the lab.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG