Principaux documents
BR146508
BRAND® glass powder funnel short wide stem
top diam. 60 mm, bottom diam. 18 mm
Synonyme(s) :
Funnels, Power funnels
1 EA
11.00 CHF
11.00 CHF
Check Cart for Availability
Changer de vue
1 EA
11.00 CHF
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
11.00 CHF
Check Cart for Availability
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 146508
Diam. du fond
18 mm
Diam. de la partie supérieure
60 mm
Description générale
The BRAND® Glass Powder Funnel with Short Wide Stem is expertly crafted from Boro 3.3 glass, ensuring durability and high performance in laboratory applications. This funnel features a ground and fire-polished upper rim, providing a smooth edge for safe handling and easy pouring. Designed with a short, wide stem, it facilitates efficient transfer of powders and granular materials, minimizing the risk of clogs and ensuring a swift flow.
Caractéristiques et avantages
  • BRAND® borosilicate glass powder funnel is best suited for transferring powders.
  • These general lab use funnels include ground and fire-polished upper rim.
  • Features a short, wide stem that facilitates optimal filtration.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG