Principaux documents
BR13933
BRAND® spare burette tubes for compact burette
size 10 mL, SILBERBRAND, amber AR-Glass
Synonyme(s) :
Accessories, Burettes, Compact Burettes, SILBERBRAND
1 EA
22.80 CHF
22.80 CHF
Date d'expédition estimée le23 mai 2025
Changer de vue
1 EA
22.80 CHF
About This Item
Code UNSPSC :
41121808
Nomenclature NACRES :
NB.43
22.80 CHF
Date d'expédition estimée le23 mai 2025
Matériaux
amber AR-Glass
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 13933
Longueur
350 mm
Taille
10 mL
Description générale
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 clear glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG