Principaux documents
BMSW1000200
Sigma-Aldrich
Accuris F1 Grade Calibration Weight
200 g
Synonyme(s) :
milligram calibration weight, top pan balance weight, top pan calibration
1 EA
103.00 CHF
103.00 CHF
Check Cart for Availability
Changer de vue
1 EA
103.00 CHF
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
103.00 CHF
Check Cart for Availability
Fabricant/nom de marque
Benchmark Scientific W1000-200
Description générale
  • F1 Grade tolerance per OIML (International Organization of Legal Metrology)
  • Works with all Accuris Lab balances
  • Non-Magnetic Stainless Steel
  • Includes a protective plastic case
Informations légales
Accuris is a trademark of Benchmark Scientific, Inc.