Principaux documents
BAH500250009
SP Bel-Art® Glove Box Iris Port Entry Cover
includes foam gasket material to create tight seal
Synonyme(s) :
glove box
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Bel-Art H50025-0009
Description générale
Iris port cover works with H50025-0020 Glove Box Iris Port Entry.
Caractéristiques et avantages
  • Allows iris ports to be closed off when glove box is not in use
  • Includes foam gasket material to create tight seal
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP