Principaux documents
BAH367180000
SP Bel-Art® Quaverette Micro-Blade Vibrating Spatula
stainless steel blade
Synonyme(s) :
spoon
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
stainless steel blade
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art H36718-0000
Catégories apparentées
Description générale
Make accurate additions of precious materials as needed. Rotating the toothed thumb wheel vibrates the curved tip spatula, giving you control over transfer from spatula to weighing dish.
Caractéristiques et avantages
Available with a plain stainless steel blade or with a brown FEP coating
Coating complies with FDA regulations governing direct food contact applications; reduces transfer losses
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP