Skip to Content
Merck
All Photos(2)

Key Documents

V900724

Sigma-Aldrich

1,3,5-Trimethoxybenzene

Vetec, reagent grade, 98%

Synonym(s):

Phloroglucinol trimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OCH3)3
CAS Number:
Molecular Weight:
168.19
Beilstein:
1307993
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

98%

bp

255 °C (lit.)

mp

50-53 °C (lit.)

SMILES string

COc1cc(OC)cc(OC)c1

InChI

1S/C9H12O3/c1-10-7-4-8(11-2)6-9(5-7)12-3/h4-6H,1-3H3

InChI key

LKUDPHPHKOZXCD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Spasfon].
P Lange
Soins. Gynecologie, obstetrique, puericulture, pediatrie, (18)(18), 43-43 (1982-11-01)
Dongqiang Zhu et al.
Environmental science & technology, 39(11), 3990-3998 (2005-06-30)
Environmental black carbon (BC) is believed to be an important adsorbent of organic pollutants. In this study, we examined the effects of changes in surface properties and adsorbate structure. A series of apolar compounds (cyclohexane, 1,2-dichlorobenzene, 1,4-xylene, 1,2,3,5-tetramethylbenzene, 1,3,5-triethylbenzene) and
A N Mayeno et al.
The Journal of biological chemistry, 264(10), 5660-5668 (1989-04-05)
Human eosinophils preferentially utilize bromide to generate a brominating agent, even at physiological halide concentrations, where chloride (140 mM) is over 1000-fold greater than bromide (20-100 microM). Under the same conditions, neutrophils use chloride to generate a chlorinating agent. The
[Effects of liver disease and age on metabolism of trimethoxybenzene (author's transl)].
P Allain et al.
Therapie, 35(5), 591-595 (1980-09-01)
Xichen Cai et al.
The journal of physical chemistry. A, 111(22), 4743-4747 (2007-05-08)
Bimolecular hole transfer quenching of the 1,3,5-trimethoxybenzene radical cation (TMB*+) in the excited state (TMB*+*) by hole quenchers (Q) such as biphenyl (Bp), naphthalene (Np), anisole (An), and benzene (Bz) with higher oxidation potentials than that of TMB was directly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service