Skip to Content
Merck
All Photos(1)

Key Documents

8.18887

Sigma-Aldrich

Phloroglucinol

for synthesis

Synonym(s):

Phloroglucinol, 1,3,5-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 114.00

CHF 114.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
CHF 114.00

About This Item

Linear Formula:
C6H3-1,3,5-(OH)3
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-611-2
NACRES:
NA.22

CHF 114.00


Estimated to ship on23 May 2025


Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

solid

potency

4000 mg/kg LD50, oral (Rat)

mp

217-220 °C

solubility

10 g/L

bulk density

560 kg/m3

storage temp.

2-30°C

SMILES string

Oc1cc(cc(c1)O)O

InChI

1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H

InChI key

QCDYQQDYXPDABM-UHFFFAOYSA-N

Application

  • Phloroglucinol in α-glycosidase inhibition: Phloroglucinol derivatives demonstrate effective α-glycosidase inhibitory activity, indicating potential for application in managing diabetes through enzyme inhibition strategies (Sevimli et al., 2024).
  • Phenolic compound bioaccessibility studies: Investigating the intestinal absorption and bioaccessibility of phloroglucinol, among other phenolics, using in vitro simulated gastrointestinal digestion helps understand its pharmacokinetic properties in dietary and pharmaceutical contexts (Pais et al., 2024).
  • Applications in plant biotechnology: Phloroglucinol is utilized in Agrobacterium-mediated transformations, enhancing genetic modification techniques in plant sciences, specifically demonstrated in dwarf soybean transformations (Shao et al., 2024).
  • Methane inhibition studies in dairy cows: Phloroglucinol′s role as a hydrogen acceptor in rumen pulse-dosing experiments demonstrates its potential in reducing methane emissions from dairy cows, contributing to environmental sustainability in agricultural practices (Maigaard et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service