Skip to Content
Merck
All Photos(1)

Key Documents

442492

Supelco

Bis(2-ethylhexyl) adipate

analytical standard

Synonym(s):

Adipic acid di(2-ethylhexyl) ester, DOA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
Beilstein:
1803774
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 250 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.447 (lit.)

bp

175 °C/2 mmHg (lit.)

mp

<−70 °C (lit.)

density

0.925 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

InChI

1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3

InChI key

SAOKZLXYCUGLFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirsten Jarfelt et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 505-515 (2005-03-08)
Di(2-ethylhexyl) phthalate (DEHP) is a well-known testicular toxicant inducing adverse effects in androgen responsive tissues. Therefore, di(2-ethylhexyl) adipate (DEHA) is currently being evaluated as a potential substitute for DEHP. Similarities in structure and metabolism of DEHP and DEHA have led
Yadollah Yamini et al.
Journal of separation science, 32(18), 3201-3208 (2009-09-12)
Two approaches based on solidification of floating drop microextraction (SFDME) and homogenous liquid-liquid microextraction (HLLE) were compared for the extraction and preconcentration of di-(2-ethylhexyl) phthalate (DEHP) and di-(2-ethylhexyl) adipate (DEHA) from the mineral water samples. In SFDME, a floated drop
Kyoko Nabae et al.
Reproductive toxicology (Elmsford, N.Y.), 22(3), 411-417 (2006-09-06)
F344 male rats were given five consecutive weekly subcutaneous injections of folic acid for induction of chronic renal dysfunction and then di(2-ethylhexyl)phthalate (DEHP) or di(2-ethylhexyl)adipate (DEHA) in the diet at a concentration of 0, 6000 or 25,000 ppm for 4
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 19(4), 517-525 (2005-03-08)
This study aimed to characterize the effects of di(2-ethylhexyl) phthalate (DEHP) on the fetal rat testes and relate them to the effects seen in adults. Histopathological effects in fetal testes were examined with immunohistochemistry for anti-Mullerian hormone (AMH), 3beta-hydroxysteroid dehydrogenase
Julie Borch et al.
Reproductive toxicology (Elmsford, N.Y.), 18(1), 53-61 (2004-03-12)
The plasticizer di(2-ethylhexyl)phthalate (DEHP) exhibits antiandrogenic effects in perinatally exposed male rats. Di(2-ethylhexyl) adipate (DEHA) and diisononyl phthalate (DINP) are currently being evaluated as potential substitutes for DEHP, but similarities in structure and metabolism of DEHP with DEHA and DINP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service