Merck
  • Toxicity of di(2-ethylhexyl)phthalate (DEHP) and di(2-ethylhexyl)adipate (DEHA) under conditions of renal dysfunction induced with folic acid in rats: enhancement of male reproductive toxicity of DEHP is associated with an increase of the mono-derivative.
Toxicity of di(2-ethylhexyl)phthalate (DEHP) and di(2-ethylhexyl)adipate (DEHA) under conditions of renal dysfunction induced with folic acid in rats: enhancement of male reproductive toxicity of DEHP is associated with an increase of the mono-derivative.
Reproductive toxicology (Elmsford, N.Y.) (2006-09-06)
Kyoko Nabae, Yuko Doi, Satoru Takahashi, Toshio Ichihara, Chitose Toda, Koji Ueda, Yoshinori Okamoto, Nakao Kojima, Seiko Tamano, Tomoyuki Shirai
ABSTRACT
F344 male rats were given five consecutive weekly subcutaneous injections of folic acid for induction of chronic renal dysfunction and then di(2-ethylhexyl)phthalate (DEHP) or di(2-ethylhexyl)adipate (DEHA) in the diet at a concentration of 0, 6000 or 25,000 ppm for 4 weeks in order to investigate whether male reproductive toxicity of the two chemicals might be enhanced under conditions of renal disease. Control animals also received DEHP or DEHA in the same manner but without folic acid pretreatment. Decreased testicular weights, seminiferous atrophy with vacuolization of sertoli cells and diminished sperm counts were more prominent in rats given folic acid and then 25,000 ppm DEHP as compared to those exposed to DEHP alone. No such reproductive toxicity was evident in rats given 6000 ppm DEHP or either dose of DEHA. An increased concentration of the mono-derivative of DEHP (mono(2-ethylhexyl)phthalate, MEHP) in the blood, testis and urine was considered relevant to the enhanced reproductive toxicity observed with DEHP.
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥97.0% (GC)
Sigma-Aldrich
Bis(2-ethylhexyl) adipate, ≥99%
Supelco
Di(2-ethylhexyl)adipate solution, certified reference material, 2000 μg/mL in methanol
Supelco
Bis(2-ethylhexyl) adipate, Selectophore, for ion-selective electrodes, ≥99.0%
Supelco
Bis(2-ethylhexyl) adipate, 99%