About This Item
grade
analytical standard
packaging
ampule of 1000 mg
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
bp
255-256 °C/740 mmHg (lit.)
mp
53-56 °C (lit.)
application(s)
environmental
format
neat
storage temp.
room temp
SMILES string
Oc1c(Br)cccc1Br
InChI
1S/C6H4Br2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H
InChI key
SSIZLKDLDKIHEV-UHFFFAOYSA-N
Gene Information
human ... ALOX12(239) , ALOX15(246) , GABRA1(2554)
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service