Z708453
One Well rack
Epoxy coated wire, yellow, Holds 1 x 50 mL tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
metal
feature
Holds 1 x 50 mL tube
manufacturer/tradename
Heathrow Scientific HS23051
technique(s)
cryopreservation: suitable
size
90x67x67 mm
color
yellow
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Move the wire and friction-fit rack to exactly where you need your tube. Friction-fit racks securely hold conical or round bottom tubes with 27.5 to 29 mm OD. Link the interlocking plastic racks to form a line of 50 mL tubes. Both the wire and interlocking polypropylene racks are autoclavable. Interlocking racks are sold in assorted colors of blue, green, yellow, pink, and orange.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service