EP0030119860
Eppendorf® Single Tube Stand for Conical Tubes 25 mL and 50 mL
case of 4 pieces, white, autoclavable
Select a Size
CHF 26.40
Select a Size
About This Item
CHF 26.40
Recommended Products
material
white
feature
autoclavable
packaging
case of 1 ea
case of 4 pieces
manufacturer/tradename
Eppendorf® 0030119860
Related Categories
General description
- Stackable within one rack format and also between all rack variants?–?saves laboratory space when not in use
- Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
- Wavelike design: Tube rows are arranged in different heights so that the reaction vessels are easily accessible and clearly identifiable
- Made of polypropylene (PP) for high chemical and physical resistance
- Optimized space-saving dimensions due to perfect tube position-to-footprint arrangement without compromising the tube handling
- High flexibility: Different sized tubes fit in the 1.5/2.0 mL, 5.0/15.0 mL, 5.0/15/50 mL, and cryogenic tube rack
- Numbered tube positions support fast and easy orientation and error-free handling
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)