Skip to Content
Merck
All Photos(2)

Documents

SRP6047

Sigma-Aldrich

TNF receptor-associated protein 1 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

TNFR-associated protein 1, TRAP 1, TRAP-1, TRAP1, Tumor necrosis factor type 1 receptor-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

76.5 kDa

packaging

pkg of 10 μg

concentration

~13.5 mg/mL

impurities

Endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TRAP1(10131)

General description

Human TRAP1 belongs to the mitochondrial heat shock protein family. These molecular chaperones are highly conserved and play an important role in signal transduction, protein folding and degradation. Recombinant human TRAP1 has a C-terminal FLAG tag and has 702 amino acid residues. It has been identified to be protective for cell survival, and has been suggested as a target for anti-cancer therapeutics. Also can be useful for studies including enzyme kinetics, activator screening and selectivity profiling.

Physical form

50 mM potassium phosphate pH 7.4, 50 mM sodium chloride, 0.5 mM DTT, 0.5 mM EDTA, and 2.5% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service