Skip to Content
Merck
All Photos(1)

Documents

TT

92050721, human thyroid, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TT, 92050721

biological source

human thyroid

growth mode

Adherent

karyotype

Aneuploid karyotype with marker chromosomes

morphology

Not specified

products

Calcitonin, Carcinoembryonic antigen (CEA)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian thyroid medullary carcinoma

Cell Line Description

Established from a thyroid medullary carcinoma of a 77 year old Caucasian female. Tumourigenic in nude mice.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,13
D13S317: 11
D16S539: 12,13
D5S818: 12,13
D7S820: 10,12
THO1: 6,9
TPOX: 8,11
vWA: 16,18

Culture Medium

Ham′s F12 + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 1% Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Population doubling is very slow, cells can take up to 5 days to reach 50% confluency; media change 2-3 times a week is recommen

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service