Skip to Content
Merck
All Photos(1)

Key Documents

PZ0131

Sigma-Aldrich

PF-998425

≥98% (HPLC)

Synonym(s):

4-((1R,2R)-2-Hydroxycyclohexyl)-2(trifluoromethyl)benzonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CHF 148.00
25 MG
CHF 514.00

CHF 148.00


Estimated to ship on05 May 2025



Select a Size

Change View
5 MG
CHF 148.00
25 MG
CHF 514.00

About This Item

Empirical Formula (Hill Notation):
C14H14F3NO
CAS Number:
Molecular Weight:
269.26
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

CHF 148.00


Estimated to ship on05 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

room temp

SMILES string

O[C@@H]1CCCC[C@@H]1c2ccc(C#N)c(c2)C(F)(F)F

InChI

1S/C14H14F3NO/c15-14(16,17)12-7-9(5-6-10(12)8-18)11-3-1-2-4-13(11)19/h5-7,11,13,19H,1-4H2/t11-,13-/m1/s1

InChI key

MENRRRXHFQYXDW-DGCLKSJQSA-N

Biochem/physiol Actions

PF-998425 is a novel, nonsteroidal androgen receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to Nuclear Receptors (Steroids) for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service