118510
ATR Kinase Inhibitor II
Synonym(s):
ATR Kinase Inhibitor II, 3-Amino-6-(4-((1-(dimethylamino)propan-2-yl)sulfonyl)-phenyl)-N-phenylpyrazine-2-carboxamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥97% (HPLC)
Quality Level
form
powder
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
yellow
solubility
DMSO: 100 mg/mL
shipped in
wet ice
storage temp.
2-8°C
General description
A cell-permeable aminopyrazinyl-sulfone compound that acts as a potent, reversible and ATP-competitive inhibitor of ATR kinase (IC50 = 12 nM; Ki = 6 nM) with excellent selectivity over ATM, DNA-PK (IC50 >8 µM) and in a 50-kinase panel (% inhibition at 2 µM = 59, 57, 55, 52, 43 and 42 for AMPK, Lck, RIPK2, Ret, IKKα and FGFR3, respectively). Shown to block pH2AX-Ser139 (IC50 = 420 nM in hydroxyurea treated HT29 cells) and preferentially enhance cisplatin (Cat. No. 232120) cytotoxicity against tumor cells over normal cells by ~7-fold at 620 nM.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Other Notes
Charrier, J.D., et al. 2011. J. Med. Chem.54, 2320.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service