P3738
Protein Kinase G Iβ human
≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf9 cells, buffered aqueous glycerol solution
Synonym(s):
cyclic-Guanosine Monophosphate-dependent Protein Kinase 1β human
About This Item
Recommended Products
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
Assay
≥95% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
≥1.5 units/mg protein (20-fold stimulation by cGMP (5 μM))
mol wt
76 kDa (monomer)
UniProt accession no.
relevant disease(s)
cancer
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... PRKG1(5592)
Application
Biochem/physiol Actions
Unit Definition
Physical form
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service