Skip to Content
Merck
All Photos(1)

Documents

P3591

Sigma-Aldrich

1,2-Dioctanoyl-sn-glycerol 3-phosphate sodium salt

≥99%

Synonym(s):

L-α-Phosphatidic acid, dioctanoyl sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H36O8PNa
CAS Number:
Molecular Weight:
446.45
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥99%

form

powder

storage temp.

−20°C

InChI

1S/C19H37O8P.Na/c1-3-5-7-9-11-13-18(20)25-15-17(16-26-28(22,23)24)27-19(21)14-12-10-8-6-4-2;/h17H,3-16H2,1-2H3,(H2,22,23,24);/q;+1/p-1/t17-;/m1./s1

InChI key

ZNDNSOYDPUTHKQ-UNTBIKODSA-M

General description

1,2-Dioctanoyl-sn-glycerol 3-phosphate sodium salt is a cell-permeable phosphatidic acid (PA).

Application

1,2-Dioctanoyl-sn-glycerol 3-phosphate sodium salt has been used:
  • to study its effects on neural stem/precursor cell (NSC) proliferation
  • to study the effects of phosphatidic acid (PA) on actin filament dynamics in Arabidopsis thaliana epidermal leaf cells
  • to treat vector-transfected cells and to verify the attenuation of hypoxia-inducible factor-1α (HIF-1α) induction in phospholipase D (PLD)-transfected cells by butanol

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ETOH inhibits embryonic neural stem/precursor cell proliferation via PLD signaling
Fujita Y, et al.
Biochemical and Biophysical Research Communications, 370(1), 169-173 (2008)
Jindřiška Matoušková et al.
Plant science : an international journal of experimental plant biology, 223, 36-44 (2014-04-29)
Changes in actin cytoskeleton dynamics are one of the crucial players in many physiological as well as non-physiological processes in plant cells. Positioning of actin filament arrays is necessary for successful establishment of primary lines of defense toward pathogen attack
Yuko Fujita et al.
Biochemical and biophysical research communications, 370(1), 169-173 (2008-03-25)
While a mother's excessive alcohol consumption during pregnancy is known to have adverse effects on fetal neural development, little is known about the underlying mechanism of these effects. In order to investigate these mechanisms, we investigated the toxic effect of
Phospholipase D activates HIF-1-VEGF pathway via phosphatidic acid
Han S, et al.
Experimental & Molecular Medicine, 46(12), e126-e126 (2014)
Songyi Han et al.
Experimental & molecular medicine, 46, e126-e126 (2014-12-20)
Growth factor-stimulated phospholipase D (PLD) catalyzes the hydrolysis of phosphatidylcholine (PC), generating phosphatidic acid (PA) which may act as a second messenger during cell proliferation and survival. Therefore, PLD is believed to play an important role in tumorigenesis. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service