Skip to Content
Merck
All Photos(1)

Documents

I7015

Sigma-Aldrich

D-myo-Inositol 1,2,5,6-tetra­kis­phos­phate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16O18P4
CAS Number:
Molecular Weight:
500.08
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

N.O[C@@H]1[C@@H](O)[C@@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H16O18P4.H3N/c7-1-2(8)4(22-26(12,13)14)6(24-28(18,19)20)5(23-27(15,16)17)3(1)21-25(9,10)11;/h1-8H,(H2,9,10,11)(H2,12,13,14)(H2,15,16,17)(H2,18,19,20);1H3/t1-,2-,3-,4+,5-,6-;/m1./s1

InChI key

MPALWCVIGLCHDH-TYZYTZLSSA-N

Application

Used to study the hormonal control of Ca2+-dependent processes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Q Phillippy et al.
Analytical biochemistry, 175(1), 162-166 (1988-11-15)
Inositol phosphates including phytic acid were separated in 30 min by gradient ion chromatography with postcolumn derivatization. All four pentakisphosphates were resolved, while four tetrakisphosphate peaks were detected. The limits of detection for all polyphosphates, including tris- and bisphosphates, were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service