Skip to Content
Merck
All Photos(1)

Key Documents

M0699

Sigma-Aldrich

MSK1, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC1911, MSPK1, RLPK, RPS6KA5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 470.00

CHF 470.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 470.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

CHF 470.00


Estimated to ship on29 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

75-103 nmol/min·mg

mol wt

~120 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RPS6KA5(9252)

Biochem/physiol Actions

MSK1 or mitogen- and stress-activated protein kinase-1 contains 2 protein kinase domains and shares 43% protein sequence identity with the MAPKAPK1 isoforms. Northern blot analysis shows that MSK1 is expressed in all tissues with the highest level of expression in brain, muscle, and placenta. Immunoelectron microscopy localized MSK1 to the nucleus. MSK1 is activated in vitro and in vivo by either ERK or SAPK2 proteins. MSK1 rather than MAPKAP-K1 or MAPKAP-K2/K3, mediates activation of the cAMP response element-binding protein and activating transcription factor-1 by either growth factors or stress signals. By radiation hybrid analysis, the RPS6KA5 gene has been mapped to chromosome 14q31-q32.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of a member of the ribosomal protein S6 kinase family, RPS6KA5, to human chromosome 14q31-->q32.1 by radiation hybrid mapping.
C Jiang et al.
Cytogenetics and cell genetics, 87(3-4), 261-262 (2000-03-07)
M Deak et al.
The EMBO journal, 17(15), 4426-4441 (1998-08-04)
We have identified a novel mitogen- and stress-activated protein kinase (MSK1) that contains two protein kinase domains in a single polypeptide. MSK1 is activated in vitro by MAPK2/ERK2 or SAPK2/p38. Endogenous MSK1 is activated in 293 cells by either growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service