Skip to Content
Merck
All Photos(2)

Documents

HS120882

Hexagonal weighing boats

size M, anti-static, sterile; electron beam irradiated

Synonym(s):

weighing dish, weigh boat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NA.74

material

hexagonal white polystyrene

size

M

sterility

sterile; electron beam irradiated

feature

anti-static

packaging

pack of 200 ea (40 x packs of 5 ea)

manufacturer/tradename

Heathrow Scientific 120882

L × W × H

2.5 in. (6.4 cm) × 2 in. (5.1 cm) × .7 in. (1.9 cm)

capacity

46 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Hexagonal weighing boats are ideal for handling samples for weighing.
  • Flexible boats can be easily bent to form pour spouts for spill-free transfer and prevent sample loss.
  • Flat bottoms and sloping slides provide an accurate transfer.
  • These antistatic, polystyrene boats are non-reactive to most substances.
  • Chemically resistant to most chemicals including weak acids, aqueous solutions, alcohols, and bases.
  • Compatible with microwave moisture analyzers.
  • Resist temperatures to 158 °F (70 °C).

Application

For life science or molecular research, bio-pharmaceutical, and environmental

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service