Skip to Content
Merck
All Photos(2)

Documents

HS120880

Weigh Boat

size small, square white polystyrene, anti-static, capacity 10 mL, sterile; electron beam irradiated

Synonym(s):

weighing dish, weigh boat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71

material

square white polystyrene

size

small

sterility

sterile; electron beam irradiated

feature

anti-static

packaging

pack of 200 ea (40 x packs of 5 ea)

manufacturer/tradename

Heathrow Scientific 120880

L × W × H

1.8 in. (4.6 cm) × 1.8 in. (4.6 cm) × .3 in. (.8 cm)

capacity

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.

Application

For life science or molecular research, bio-pharmaceutical, and environmental

Features and Benefits

  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service