Skip to Content
Merck
All Photos(1)

Documents

G5173

Sigma-Aldrich

Garcinol

≥95%

Synonym(s):

(1R,5R,7R)-3-(3,4-Dihydroxybenzoyl)-4-hydroxy-8,8-dimethyl-1,7-bis(3-methyl-2-buten-1-yl)-5-[(2S)-5-methyl-2-(1-methylethenyl)-4-hexen-1-yl]-Bicyclo[3.3.1]non-3-ene-2,9-dione, Camboginol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H50O6
CAS Number:
Molecular Weight:
602.80
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95%

form

powder

color

yellow

solubility

>5 mg/mL

storage temp.

−20°C

SMILES string

C\C(C)=C\C[C@@H](C[C@@]12C[C@@H](C\C=C(\C)C)C(C)(C)[C@@](C\C=C(/C)C)(C(=O)C(=C1O)C(=O)c3ccc(O)c(O)c3)C2=O)C(C)=C

InChI

1S/C38H50O6/c1-22(2)11-13-27(25(7)8)20-37-21-28(15-12-23(3)4)36(9,10)38(35(37)44,18-17-24(5)6)34(43)31(33(37)42)32(41)26-14-16-29(39)30(40)19-26/h11-12,14,16-17,19,27-28,39-40,42H,7,13,15,18,20-21H2,1-6,8-10H3/t27-,28+,37+,38-/m0/s1

InChI key

QDKLRKZQSOQWJQ-SMDXAGPFSA-N

Biochem/physiol Actions

Garcinol is a HAT (histone acetyltransferase) inhibitor. Garcinol is anti-inflammatory and anti-carcinogenic. Histone acetyltransferases provide opposing enzymatic mechanisms for chromatin remodeling and epigenetic control of gene expression. HATs are involved in regulation of transformation, transcription, differentiation, apoptosis, embryonic development and oncogenesis. IC50 p300 = 7 mM; IC50 PCAF = 5 mM (similar to anacardic acid). Garcinol is highly toxic compared to LTK-14, and does not discriminate between p300 and PCAF.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aamir Ahmad et al.
Molecular cancer therapeutics, 11(10), 2193-2201 (2012-07-24)
Anticancer properties of Garcinia indica-derived garcinol are just beginning to be elucidated. We have earlier reported its cancer cell-specific induction of apoptosis in breast cancer cells, which was mediated through the downregulation of NF-κB signaling pathway. To gain further mechanistic
Zaorui Zhao et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(7), 2344-2351 (2012-03-08)
Histone acetylation has recently been implicated in learning and memory processes, yet necessity of histone acetylation for such processes has not been demonstrated using pharmacological inhibitors of histone acetyltransferases (HATs). As such, the present study tested whether garcinol, a potent
Aamir Ahmad et al.
Carcinogenesis, 33(12), 2450-2456 (2012-09-14)
Garcinol, obtained from Garcinia indica, has exhibited some promising anticancer activity. In particular, our earlier work has demonstrated its ability to inhibit cell proliferation and induction of apoptosis in multiple cancer cell lines representative of breast, prostate, as well as
Chandra Sekhar Rao Kadiyala et al.
The Journal of biological chemistry, 287(31), 25869-25880 (2012-06-01)
Histone acetylation was significantly increased in retinas from diabetic rats, and this acetylation was inhibited in diabetics treated with minocycline, a drug known to inhibit early diabetic retinopathy in animals. Histone acetylation and expression of inflammatory proteins that have been
Chih-Hsiung Wu et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 17(11), 3533-3541 (2011-03-30)
The nicotinic acetylcholine receptor (nAChR) was first characterized in 1970 as a membrane receptor of a neurotransmitter and an ion channel. nAChRs have been shown to be involved in smoking-induced cancer formation in multiple types of human cancer cells. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service