Merck
  • Nicotinic acetylcholine receptor-based blockade: applications of molecular targets for cancer therapy.
Nicotinic acetylcholine receptor-based blockade: applications of molecular targets for cancer therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2011-03-30)
Chih-Hsiung Wu, Chia-Hwa Lee, Yuan-Soon Ho
ABSTRACT
The nicotinic acetylcholine receptor (nAChR) was first characterized in 1970 as a membrane receptor of a neurotransmitter and an ion channel. nAChRs have been shown to be involved in smoking-induced cancer formation in multiple types of human cancer cells. In vitro and in vivo animal studies have shown that homopentameric nAChR inhibitors, such as methyllycaconitine and α-Bgtx, can attenuate nicotine-induced proliferative, angiogenic, and metastatic effects in lung, colon, and bladder cancer cells. Recent publications have shown that α9-nAChR is important for breast cancer formation, and in many in vivo studies, α9-nAChR-specific antagonists (e.g., α-ImI, α-ImI, Vc1.1, RgIA, and It14a) produced an analgesic effect. Vc1.1 functions in a variety of animal pain models and currently has entered phase II clinical trials. For cancer therapy, natural compounds such as garcinol and EGCG have been found to block nicotine- and estrogen-induced breast cancer cell proliferation through inhibition of the α9-nAChR signaling pathway. A detailed investigation of the carcinogenic effects of nAChRs and their specific antagonists would enhance our understanding of their value as targets for clinical translation.