Documents
EPM1192-9901
Quick-change platform kit for Eppendorf® New Brunswick Innova® 2100, 4000, 4080, 4230 shaker
enables platform attachment without tools
Synonym(s):
Quick Change Platform Kit
About This Item
UNSPSC Code:
41103816
NACRES:
NB.73
feature
enables platform attachment without tools
manufacturer/tradename
Eppendorf® M1192-9901
Related Categories
General description
The Eppendorf M1192-9901 quick-change platform accessory kit converts Eppendorf shakers to allow interchangeable platforms to be installed without tools. It is for use with the Eppendorf New Brunswick Innova 2100 series platform shaker, Excella E24/E24R series incubator shaker, and Innova 40/40R and 42/42R series incubator shaker (all sold separately). The kit includes a subplatform with spring clips, counterweight, and installation hardware.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG