Skip to Content
Merck
All Photos(2)

Documents

EP951020320

Eppendorf twin.tec® PCR Plates 96

semi skirted, maximum volume 250 μL, yellow, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300

sterility

non-sterile

feature

skirt
semi skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020320

maximum volume

250 μL

size

96 well

working volume

-200 μL

color

yellow

suitability

suitable for PCR
suitable for molecular biology

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf “twin.tec” PCR Plates 96 are 96 well PCR plates used extensively in various polymerase chain reaction (PCR) applications. These polypropylene plates provide excellent thermal conductivity, ensuring uniform temperature distribution during PCR.

Application

Eppendorf “twin.tec” PCR Plates 96 may be used in a wide variety of applications such as:
  • Standard PCR
  • Real time PCR
  • High throughput PCR

PCR

Features and Benefits

  • The Eppendorf twin.tec PCR Plates are made from sturdy polycarbonate frames, which help in maintaining the shape at elevated temperatures.
  • They have raised rims which ensures effective sealing on the plates, preventing cross contamination.
  • Eppendorf [TM=“twin.tec”] PCR Plates 96 are semi-skirted with a maximum well volume of 250 µL.
  • These plates are compatible with most thermal cyclers.
  • Twin.tec plates are ideal for quantitative real time PCR.
  • Certified free of human DNA, DNase, RNase, and PCR inhibitors.
  • Eppendorf twin.tec PCR Plates display improved well – well tolerance.
  • They are semi-skirted for the purpose of labeling and are available with barcodes, upon request.
  • The [TM=“OptiTrack”] matrix feature enables faster sample identification and reduces pipetting errors.

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service