Key Documents
EP5804756002
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer, Falcon and micro test tubes 7-18 mL, pack of 2 ea
Select a Size
2 EA
CHF 169.00
CHF 169.00
Check Cart for Availability
Select a Size
Change View
2 EA
CHF 169.00
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
CHF 169.00
Check Cart for Availability
feature
holds standard, Vacutainer, Falcon and micro test tubes 7-18 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804756002
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL rectangular bucket
General description
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)