Key Documents
EP0030627300
Eppendorf® Lobind Microplate 384/V-PP, 384 wells
white polypropylene, V-bottom clear wells, PCR clean, pkg of 240 ea
Select a Size
240 EA
CHF 1’810.00
CHF 1’810.00
Check Cart for Availability
Select a Size
Change View
240 EA
CHF 1’810.00
About This Item
UNSPSC Code:
41122107
NACRES:
NB.15
material:
V-bottom clear wells
white polypropylene
size:
384 wells
sterility:
non-sterile
binding type:
Protein low binding
CHF 1’810.00
Check Cart for Availability
material
V-bottom clear wells
white polypropylene
sterility
non-sterile
packaging
pkg of 240 ea
manufacturer/tradename
Eppendorf® 0030627300
technique(s)
PCR: suitable
maximum volume
-140 μL
size
384 wells
working volume
5-120 μL
binding type
Protein low binding
General description
Maximize your sample recovery with Eppendorf LoBind Plates. Eppendorf LoBind Plates are designed with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for research or sensitive testing methods where they enable excellent results due to less sample binding to the plate surface.
The “Eppendorf” Lobind Microplate 384/V-PP is a DNA LoBind plate having 384 wells. It improves the recovery of nucleic acids by reducing their adsorption to the walls of the tube. Special manufacturing techniques along with selected polypropylene batches ensure nearly 100?% recovery of DNA/RNA molecules-without surface coating, thereby eliminating the risk of sample contamination.
Application
The “Eppendorf” Lobind Microplate 384/V-PP is suitable for:
  • Preparation or storage of DNA and RNA samples
  • Forensic trace analysis
  • Preparing dilution series in quantitative real-time PCR
  • Sample preparation in next-generation sequencing
  • Creation of genomic or oligonucleotide libraries
Features and Benefits
  • The Lobind feature ensures maximum sample recovery thereby improving assay results.
  • The [TM=“Eppendorf”] Lobind Microplate 384/V-PP is devoid of surface coatings to minimize sample interference.
  • Raised well rims and a smooth surface ensure a secure sealing in plates.
  • An optimized [TM=“RecoverMax”] well geometry minimizes the remaining volume and ensures smooth mixing properties.
  • The distinctive [TM=“OptiTrack”] matrix offers accelerated sample identification and reduces pipetting errors.
  • Free of human DNA, DNase, RNase, and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)