Skip to Content
Merck
All Photos(1)

Documents

EP0030129512

Eppendorf twin.tec® PCR Plates 96

Skirted, LoBind, PCR clean, working volume 150 μL, colorless, pack of 25

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.15

material

colorless

well size

96

sterility

non-sterile

feature

skirt
LoBind
PCR clean
Skirted

packaging

pack of 25

manufacturer/tradename

Eppendorf® 0030129512 (Global)

technique(s)

PCR: suitable

size

96 well

working volume

150 μL

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf twin.tec® 96 Well PCR Plates improve recovery of nucleic acids by reducing their absorption to the tube wall. A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100 % recovery of DNA/RNA molecules without surface coating to eliminate the risk of sample contamination.Batch-tested and certified by an independent laboratory to be free from DNA, DNase, RNase, and PCR inhibitors.

Application

  • PCR with low template concentration
  • Real-time PCR with low sample concentration
  • Low volume PCR/qPCR
  • NGS DNA library construction
PCR

Features and Benefits

  • Maximum recovery rates for improved assay results.
  • One-piece design: Combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Exceptionally solid and torque-resistant polycarbonate frame for ultimate rigidity.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample. They are designed to maximize the yield of your target molecules. DNA is less likely to bind to polypropylene and thus remains within the liquid of your reaction. Subsequently, more molecules are available for the chemical reaction, e.g. PCR.
  • The OptiTrack® matrix enables faster sample identification and minimizes pipetting errors.
  • Raised well rims provide effective sealing and reduce the risk of cross-contamination.
  • Ideal for quantitative real-time PCR as well.
  • Autoclavable (121 °C, 20 min).
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service