Key Documents
EP0030129610
Eppendorf twin.tec® PCR Plates 96
Semi-Skirted, Forensic DNA Grade, working volume 250 μL, colorless, pack of 10 (individually wrapped)
Select a Size
10 EA
CHF 82.50
CHF 82.50
Estimated to ship on20 April 2025
Select a Size
Change View
10 EA
CHF 82.50
About This Item
UNSPSC Code:
41106300
NACRES:
NB.22
material:
colorless
colorless wells
size:
96 well
working volume:
250 μL
feature:
skirt
Forensic DNA Grade
Semi-Skirted
Skirt (Semi skirted)
CHF 82.50
Estimated to ship on20 April 2025
material
colorless
colorless wells
well size
96
feature
skirt
Forensic DNA Grade
Semi-Skirted
Skirt (Semi skirted)
packaging
pack of 10 (individually wrapped)
manufacturer/tradename
Eppendorf® 0030129610 (Global)
technique(s)
PCR: suitable
size
96 well
working volume
250 μL
General description
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
Application
PCR
Features and Benefits
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)