Skip to Content
Merck
All Photos(2)

Documents

EP0030125193

Sigma-Aldrich

Eppendorf® Microtube 3810X

capacity 1.5 mL, red, pkg of 1000 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.02

material

cap (push fit)
polypropylene
red

sterility

non-sterile

packaging

pkg of 1000 ea

manufacturer/tradename

Eppendorf® 0030125193

capacity

1.5 mL

maximum volume

1.5 mL

suitability

suitable for (g-safe® centrifugation stability)

Looking for similar products? Visit Product Comparison Guide

General description

The easy to open and close design of the Eppendorf Tubes® 3810X offers convenient, ergonomic and reliable one-hand operation. Whether sample preparation, centrifugation or storage - the Eppendorf Tubes® 3810X fit perfectly into your workflows!

Features and Benefits

  • Tube lid is easy to open and close for ergonomic operation
  • Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes g-Safe®: Exceptional centrifugation stability up to 30,000 x g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
  • Graduation for visual control of the pipetted volume
  • Frosted lid and surface on the side for easy labeling
  • Ensured functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service