Skip to Content
Merck
All Photos(1)

Key Documents

CLS6524

Corning® microplate sealing tape

polyethylene film, non-sterile, suitable for (for short and long term storage and PCR, 96 well reactions)

Synonym(s):

microplate sealing film, multiwell plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
CHF 96.90

CHF 96.90


Check Cart for Availability


Select a Size

Change View
100 EA
CHF 96.90

About This Item

UNSPSC Code:
41122410
NACRES:
NB.13

CHF 96.90


Check Cart for Availability

material

polyethylene film

sterility

non-sterile

packaging

case of 100 ea

manufacturer/tradename

Corning 6524

technique(s)

PCR: suitable

color

clear

suitability

suitable for (for short and long term storage and PCR, 96 well reactions)

Looking for similar products? Visit Product Comparison Guide

General description

  • Polyethylene sealing tapes for 96 well plates prevent evaporation
  • Easy application and removal for short- and long-term storage
  • Not recommended for use above 70C and not recommended for use in thermocyclers with heated-lid

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service