Skip to Content
Merck
All Photos(1)

Key Documents

EP0030127781

Eppendorf® Microplate Sealing Films & Foils

PCR film, PCR clean, autoclavable, self-adhesive, pkg of 100 ea

Synonym(s):

PCR, Plates, Sealing films

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
CHF 186.00

CHF 186.00


Check Cart for Availability


Select a Size

Change View
100 EA
CHF 186.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.15

CHF 186.00


Check Cart for Availability

sterility

non-sterile

feature

PCR clean
autoclavable
PCR film
self-adhesive

packaging

pkg of 100 ea

manufacturer/tradename

Eppendorf® 0030127781

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® PCR films and foils for airtight sealing of your PCR plates. It provides dependable and consistent sealing performance.

Application

Eppendorf® PCR microplate sealing films are ideal sealing materials that enable tight sealing of plates during thermal cycling and storage.

Features and Benefits

  • Airtight self-adhesive films prevent sample loss due to evaporation.
  • Highly chemical resistant to most of the substances in the laboratory.
  • Easy removal without leaving behind any residue.
  • Transparent sealing offers better visual clarity and helps in the monitoring of samples.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service