Skip to Content
Merck
All Photos(1)

Key Documents

CAS9NEO

Sigma-Aldrich

Cas9 Neomycin Lenti Plasmid

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CHF 244.00

CHF 244.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CHF 244.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

CHF 244.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in E. coli

Quality Level

form

liquid

packaging

vial of 50 μL

concentration

20 ng/μL in TE buffer; DNA (1μg of plasmid DNA)

application(s)

CRISPR

selection

blasticidin

shipped in

dry ice

storage temp.

−20°C

General description

This product is a neomycin resistant lenti-Cas9 plasmid for generation of lentiviral particles. Use Sigma′s lentiviral Cas9 reagents to efficiently generate stable cell lines expressing Cas9 for CRISPR genome editing. Sigma′s lenti-Cas9 plasmid is one part of a two-vector CRISPR system with individual Cas9 and gRNA expression vectors. Please see the link above to order gRNA.

Application

Functional Genomics/Screening /Target Validation/Genome Editing

Features and Benefits

Use lentiviral particles to generate cells with stable expression of Cas9 nuclease.

Components

1ug of plasmid DNA, EF1a-Cas9-2A-Neomycin Lenti

Linkage

Click here to order gRNA plasmid and virus.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service