B8907
Nalgene® bottles, style 2104
capacity 60 mL
Synonym(s):
HDPE bottles, nalgene bottle, nalgene bottles
About This Item
Recommended Products
material
polypropylene cap
polypropylene screw closure
round bottle
translucent high-density polyethylene bottle
Joint
Joints threaded neck (28)
description
Not autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2104-0002
bottle diam. × H
39 mm × 86 mm
capacity
60 mL
General description
Features and Benefits
- Nalgene® bottles are ideal for storing chemicals, materials, and specimens.
- Laboratory-grade HDPE material for reliably low leachables and extractables.
- Suitable for use in freezers (up to -100°C).
- Rigid, uniform walls offer protection against splitting or punctures.
- Bottles have excellent durability and are chemically resistant to most acids, bases, and alcohols.
- Linerless caps ensure a leak-proof seal for sample protection.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)