Skip to Content
Merck
All Photos(2)

Key Documents

AV36652

Sigma-Aldrich

Anti-MCM7 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Minichromosome maintenance complex component 7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 417.00

CHF 417.00


Estimated to ship on25 May 2025



Select a Size

Change View
100 μL
CHF 417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 417.00


Estimated to ship on25 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

81 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MCM7(4176)

Immunogen

Synthetic peptide directed towards the middle region of human MCM7

Biochem/physiol Actions

Minichromosome maintenance complex component 7 (MCM7) is one of the mini-chromosome maintenance proteins that are crucial for eukaryotic genome replication. MCM proteins are key components of the pre-replication complex, recruit replication-related proteins and form replication forks. MCM7 interacts with receptor for activated protein kinase C 1 (RACK1) and controls DNA synthesis and the entry of the cells into S phase.

Sequence

Synthetic peptide located within the following region: HRIVKMNKSEDDESGAGELTREELRQIAEEDFYEKLAASIAPEIYGHEDV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi-Yue Zhang et al.
The American journal of pathology, 182(3), 796-805 (2013-01-15)
MCM7 is one of the pivotal DNA replication licensing factors in controlling DNA synthesis and cell entry into S phase. Its expression and DNA copy number are some of the most predictive factors for the growth and behavior of human
Matthew L Bochman et al.
Microbiology and molecular biology reviews : MMBR, 73(4), 652-683 (2009-12-01)
The Mcm2-7 complex serves as the eukaryotic replicative helicase, the molecular motor that both unwinds duplex DNA and powers fork progression during DNA replication. Consistent with its central role in this process, much prior work has illustrated that Mcm2-7 loading

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service