Skip to Content
Merck
All Photos(1)

Key Documents

360562

Sigma-Aldrich

1-Octanol

ACS spectrophotometric grade, ≥99%

Synonym(s):

Alcohol C8, Capryl alcohol, Octyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7OH
CAS Number:
Molecular Weight:
130.23
Beilstein:
1697461
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

ACS spectrophotometric grade

vapor density

4.5 (vs air)

vapor pressure

0.14 mmHg ( 25 °C)

Assay

≥99%

autoignition temp.

523 °F

expl. lim.

8 %

impurities

≤0.0002 meq/g Titr. acid

evapn. residue

≤0.004%

color

APHA: ≤10

refractive index

n20/D 1.429 (lit.)

bp

196 °C (lit.)

mp

−15 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 220 nm Amax: 1.00
λ: 230 nm Amax: 0.50
λ: 240 nm Amax: 0.10
λ: 260 nm Amax: 0.05
λ: 280-400 nm Amax: 0.01

SMILES string

CCCCCCCCO

InChI

1S/C8H18O/c1-2-3-4-5-6-7-8-9/h9H,2-8H2,1H3

InChI key

KBPLFHHGFOOTCA-UHFFFAOYSA-N

Gene Information

mouse ... Chrna1(11435)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Blocks T-type Ca2+ channels

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

187.7 °F

Flash Point(C)

86.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eagleson M.
Concise Encyclopedia Chemistry, 744-744 (1994)
Mechanisms of PVDF membrane formation by immersion-precipitation in soft (1-octanol) and harsh (water) nonsolvents.
Young TH, et al.
Polymer, 40(19), 5315-5323 (1999)
Photocatalytic degradation of 1-octanol on anchored titanium oxide and on TiO2 powder catalysts.
Yamashita H, et al.
J. Catal., 158(1), 97-101 (1996)
Characterization of titanium-silicon binary oxide catalysts prepared by the sol-gel method and their photocatalytic reactivity for the liquid-phase oxidation of 1-octanol.
Yamashita H, et al.
The Journal of Physical Chemistry B, 102(30), 5870-5875 (1998)
Cristian Gobbo et al.
ACS nano, 6(12), 10684-10698 (2012-12-12)
Controlling the molecular conformation of oligomers on surfaces through noncovalent interactions symbolizes an important approach in the bottom-up patterning of surfaces with nanoscale precision. Here we report on the design, synthesis, and scanning tunneling microscopy (STM) investigation of linear oligoamides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service